Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Gloves with Padded Palm/Wrist Support

Catalog No. 19031657
Click to view available options
Size:
Small
Medium
Hand:
Left
Right
Specific
This item is not returnable. View return policy
This item is not returnable. View return policy

Elastic polyester gloves with VEP padding in the palm, extending to the attached wrist support for impact and shock protection.

  • Fingerless design for tactile feedback
  • Suede leather wrist protection provides grip abilities
  • VEP padding in the palm extends to the attached wrist support
  • Adjustable hook and loop closure
  • Helps to prevent repetitive strain injuries

Specifications

Cuff Material Suede Leather
Color Blue, Gray
Hand Specific
Material Polyester
Product Type Anti-impact Gloves
Size Small
Finger Style Fingerless
For Use With (Application) Preventing Repetative Strain Injuries (RSI)
Glove Cuff Color Gray
Glove Cuff Style Hook and Loop
Series 701-00
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.