Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Glove Liners with Wrist Support

Catalog No. 19031768
Click to view available options
Size:
Medium
Large
X-Large
Hand:
Left
Right
Specific
This item is not returnable. View return policy
This item is not returnable. View return policy

Polycotton liners with VEP padded palms should be worn inside of work gloves for additional protection. Detachable suede leather wrist support protects from impact, and helps to prevent repetitive strain injuries.

  • Recommended to be worn inside work gloves
  • VEP padding in the palm
  • Detachable suede leather wrist support
  • Protects from impact and repetitive strain injuries
  • Ideal for use with hand tools and power tools

Specifications

Cuff Material Suede Leather
Color Blue, Gray
Hand Right
Material Polycotton
Product Type Anti-Impact Glove Liners
Size Medium
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, and Maintenance
Glove Cuff Color Gray
Glove Cuff Style Hook and Loop
Series 703-00
Length (English) 8 in.
Length (Metric) 20.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.