Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Full-Finger Anti-Impact Glove Liners with Wrist Support

Catalog No. 189992204
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
Specific
This item is not returnable. View return policy
This item is not returnable. View return policy

Liners are constructed of 4-way stretch polycotton for breathability and comfort. VEP padding extends from the palm, to midway up the fingers and into the thumb webbing for impact and shock protection.

  • Stretch polycotton ensures breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm, midway up the fingers, and thumb web area protects from impact and shock
  • Grain leather wrist support prevents over flexion and over extension to help prevent repetitive strain injuries
  • Full finger offers maximum protection
  • Designed to be worn under work glove
  • Ideal for protection from hand tools and power tools in applications such as construction, assembly, manufacturing, and maintenance

Specifications

Cuff Material Grain Leather
Color Blue, Yellow
Hand Left
Material Polycotton
Product Type Anti-Impact Glove Liners
Size Small
For Use With (Application) Construction, Assembly, Manufacturing, and Maintenance
Glove Cuff Color Yellow
Series 712-02
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.