Learn More
Impacto™ Pearl Leather Series Half-Finger Anti-Impact Gloves
Shop All Impacto Protective Products Products
Description
- Nylon fabric back ensures breathability and comfort
- Palm is made of soft pearl leather for dexterity and abrasion protection
- 1/8 in. VEP padding in the palm protects from impact and shock
- Half-finger style leaves fingers open for tactile feedback and mobility
- Hook and loop wrist closure
- Protection against electric, manual, and power tools
- Ideal for applications such as driving, sports, wheelchairs, and manufacturing
Specifications
Specifications
| Color | Black, White |
| Hand | Left |
| Material | Pearl Leather, Nylon |
| Product Type | Fingerless Gloves |
| Size | Large |
| EN388 Standard | EN388:2222 |
| Glove Cuff Style | Band Top |
| Palm Type | Padded |
| Series | 401-30 |