Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Half-Finger Anti-Impact Gloves with Wrist Support

Catalog No. p-3142815
Encompass
Click to view available options
Size:
Medium
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

A nylon fabric back ensures breathability and comfort, while the attached leather wrist support protects users from repetitive strain injuries.

  • Nylon fabric back ensures breathability and comfort
  • Soft pearl leather cover for dexterity and abrasion protection
  • VEP 1/8 in. padding in the palm, fingers, and thumb web protects from impact and shock
  • Attached leather wrist support prevents over extension and over flexion to protect from repetitive strain injuries
  • Half-finger design for tactile feedback and mobility
  • Provides protection from electric, manual, and power tools
  • Use in applications such as construction, manufacturing, and automotive repairs
  • Gloves are sold in pairs

Specifications

Back Material Nylon
Cuff Material Leather
Color Black, White
Hand Specific
Material Pearl Leather
Product Type Anti-impact Gloves
Size Medium
Finger Style Half
For Use With (Application) Construction, Manufacturing, and Automotive Repairs
Glove Cuff Color White
Series 475-30
Length (English) 8 in.
Length (Metric) 20.5 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.