Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Glove Liners with VEP Padding

Catalog No. 19088386
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
Specific

Stretch polycotton fabric ensures breathability and comfort, while VEP padding protects palms from impact and shock.

  • 4-way stretch polycotton fabric to ensure breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm
  • Fingerless design for tactile feedback and mobility
  • Designed to be worn under work gloves for an additional level of protection
  • Ideal for protection from electric, manual, and power tools
  • Sold in pairs

Specifications

Certifications/Compliance (Europe) EN 420: 2003 + A1:2009
Color Blue
Hand Specific
Material Polycotton
Product Type Glove Liners
Size X-Large
EN388 Standard EN388:3231
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Style Open
Palm Type VEP Padding
Series 501-00
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.