Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IMPAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552667
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552667 Supplier Invitrogen™ Supplier No. PA552667
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVLHEKSKGK TREGAEDKMT SGDVLSNRKM FYLLKTAFPS VQINTEEHVD AADQEVILWD HKIPEDILKE VTTPKEVPAE SVTVWIDPLD ATQEYTEDLR KYVTTMVCVA VNGK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IMPAD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IMPAD1
Gene Accession No. Q9NX62
Gene Alias 1110001C20Rik; AA408880; Ac2-190; AI451589; AL022796; B230207P20; EC 3.1.3; EC 3.1.3.25; FLJ20421; Golgi 3-prime phosphoadenosine 5-prime phosphate 3-prime phosphatase; golgi-resident nucleotide phosphatase; golgi-resident PAP phosphatase; GPAPP; IMP 3; IMP3; IMP-3; IMPA3; IMPA3IMPase 3; IMPAD1; IMPase 3; inositol monophosphatase 3; inositol monophosphatase domain containing 1; inositol monophosphatase domain-containing protein 1; Inositol-1(or 4)-monophosphatase 3; Jaws; Myo-inositol monophosphatase A3; RGD1306455
Gene Symbols IMPAD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IMPAD1. Recombinant protein control fragment (Product #RP-90042).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54928
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.