Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPDH2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986911
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6911 Supplier Bioss Supplier No. BS5924R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes the rate-limiting enzyme in the de novo guanine nucleotide biosynthesis. It is thus involved in maintaining cellular guanine deoxy- and ribonucleotide pools needed for DNA and RNA synthesis. The encoded protein catalyzes the NAD-dependent oxidation of inosine-5′-monophosphate into xanthine-5′-monophosphate, which is then converted into guanosine-5′-monophosphate. This gene is up-regulated in some neoplasms, suggesting it may play a role in malignant transformation.

Specifications

Antigen IMPDH2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IMPDH2
Gene Accession No. P12268
Gene Alias HAMAP-Rule:MF_03156}; IMP (inosine 5′-monophosphate) dehydrogenase 2; IMP (inosine monophosphate) dehydrogenase 2; IMP dehydrogenase 2; IMP dehydrogenase 2 {ECO:0000255; IMP dehydrogenase type II; IMP oxireductase 2; IMPD; IMPD 2; IMPD 2 {ECO:0000255; IMPD2; IMPDH 2; IMPDH 2 {ECO:0000255; IMPDH II; Impdh2; IMPDH-II; inosine 5′ phosphate dehydrogenase 2; inosine 5-monophosphate dehydrogenase 2; inosine 5′-phosphate dehydrogenase 2; inosine monophosphate dehydrogenase 2; inosine monophosphate dehydrogenase type II; Inosine-5′-monophosphate dehydrogenase 2; inosine-5′-monophosphate dehydrogenase 2 {ECO:0000255
Gene Symbols IMPDH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IMPDH2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3615
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.