Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | IMPG1 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunohistochemistry-Frozen |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
IMPG1 Polyclonal specifically detects IMPG1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IMPG1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
GP147, interphotoreceptor matrix proteoglycan 1, IPM150, SPACR | |
IMPG1 | |
IgG | |
Affinity Purified |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunohistochemistry-Frozen | |
Polyclonal | |
Rabbit | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
3617 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DTGEYQDWVSICQQETFCLFDIGKNFSNSQEHLDLLQQRIKQRSFPDRKDEISAEKTLGEPGETIVISTDVANVSLGPFPLTPD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title