Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IMPG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565060 Supplier Invitrogen™ Supplier No. PA565060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTGEYQDWVSI CQQETFCLFD IGKNFSNSQE HLDLLQQRIK QRSFPDRKDE ISAEKTLGEP GETIVISTDV ANVSLGPFPL TPD Highest antigen sequence identity to the following orthologs - mouse 75%, rat 71%.

May interact with hyaluronan which may serve to form a basic macromolecular scaffold comprising the insoluble interphotoreceptor matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IMPG1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IMPG1
Gene Accession No. Q17R60
Gene Alias GP147; IMPG1; interphotoreceptor matrix proteoglycan 1; Interphotoreceptor matrix proteoglycan of 150 kDa; IPM150; IPM-150; sialoprotein associated with cones and rods; SPACR; VMD4
Gene Symbols IMPG1
Host Species Rabbit
Immunogen Recombinant Human IMPG1. Recombinant protein control fragment (Product #RP-105826).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.