Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$377.03 - $665.78
Specifications
| Antigen | IMPG2 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IMPG2 Polyclonal specifically detects IMPG2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IMPG2 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| 50939 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LQAHHDRSERESPFSGSSRQPDSLSSIENAVKYNPVYESHRAGCEKYEGPYPQHPFYSSASGDVIGGLSREEIRQMYESSELSREEIQERMRVLELYANDPEFAAFVR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Vision | |
| interphotoreceptor matrix proteoglycan 2, interphotoreceptor matrix proteoglycan 200, Interphotoreceptor matrix proteoglycan of 200 kDa, IPM 200, IPM200SPACRCAN, RP56, Sialoprotein associated with cones and rods proteoglycan, Spacrcan | |
| IMPG2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title