Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IMPG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564965 Supplier Invitrogen™ Supplier No. PA564965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDGGLGSGSGQ KVDLITWPWS ETSSEKSAEP LSKPWLEDDD SLLPAEIEDK KLVLVDKMDS TDQISKHSKY EHDDRSTHFP EEEPLSGPAV PIFADTAAES ASLTLPKHIS EVPGVDDYSV TKA Highest antigen sequence identity to the following orthologs - mouse 49%, rat 48%.

Chondroitin sulfate- and hyaluronan-binding proteoglycan involved in the organization of interphotoreceptor matrix; may participate in the maturation and maintenance of the light-sensitive photoreceptor outer segment. Binds heparin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IMPG2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IMPG2
Gene Accession No. Q9BZV3
Gene Alias IMPG2; Interphotoreceptor matrix proteoglycan 2; interphotoreceptor matrix proteoglycan IPM 200; interphotoreceptor matrix proteoglycan of 200 kDa; IPM 200; IPM200; PG10.2; RP56; Rsbp; Sialoprotein associated with cones and rods proteoglycan; spacrcan; VMD5
Gene Symbols IMPG2
Host Species Rabbit
Immunogen Recombinant Human IMPG2. Recombinant protein control fragment (Product #RP-105736).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50939
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.