Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IMPG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569455
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569455 Supplier Invitrogen™ Supplier No. PA569455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Chondroitin sulfate- and hyaluronan-binding proteoglycan involved in the organization of interphotoreceptor matrix; may participate in the maturation and maintenance of the light-sensitive photoreceptor outer segment. Binds heparin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IMPG2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IMPG2
Gene Accession No. Q9BZV3
Gene Alias IMPG2; Interphotoreceptor matrix proteoglycan 2; interphotoreceptor matrix proteoglycan IPM 200; interphotoreceptor matrix proteoglycan of 200 kDa; IPM 200; IPM200; PG10.2; RP56; Rsbp; Sialoprotein associated with cones and rods proteoglycan; spacrcan; VMD5
Gene Symbols IMPG2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human IMPG2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50939
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.