Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Importin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590575 Supplier Invitrogen™ Supplier No. PA590575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHKACQSCPS EPNTAALQAA LARVVPSYMQ AVNRERERQV VMAVLEALTG VLRSCGTLTL KPPGRLAELC GVLKAVLQRK TACQDTDEEE EEEDDDQAEY DAMLLEHAGE AIPALAAAAG GDSFAPFFAG FLPLLVCKTK QGCTVAEKSF AVGTLAETIQ GLGAASAQFV SRLLPVLLST AQEADPEVRS NAIFGMGVLA EHGGHPAQEH FPKLLGLLFP LLARERHDRV RDNICGALAR LLMASPTRKP EPQVLAALLH ALPLKEDLEE WVTIGRLFSF LYQSSPDQVI DVAPELLRIC SLILADNKIP PDTKAALLLL LTFLAKQHTD SFQAALGSLP VDKAQELQAV LGLS; Positive Samples: HeLa, SH-SY5Y, A-549, HepG2, Mouse brain, Mouse lung, Rat brain; Cellular Location: Cytoplasm, Nucleus.

Importin-4 is a protein that in humans is encoded by the IPO4 gene. Functions in nuclear protein import as nuclear transport receptor. Serves as receptor for nuclear localization signals (NLS) in cargo substrates. Is thought to mediate docking of the importin/substrate complex to the nuclear pore complex (NPC) through binding to nucleoporin and the complex is subsequently translocated through the pore by an energy requiring, Ran-dependent mechanism. At the nucleoplasmic side of the NPC, Ran binds to the importin, the importin/substrate complex dissociates and importin is re-exported from the nucleus to the cytoplasm where GTP hydrolysis releases Ran.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Importin 4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ipo4
Gene Accession No. Q8TEX9, Q8VI75
Gene Alias 8430408O15Rik; AA409693; Imp4; Imp4a; IMP4B; importin 4; importin 4a; importin-4; Importin-4a; importin-4b; IPO4; Ran-binding protein 4; ranBP4
Gene Symbols Ipo4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 728-1081 of human IPO4 (NP_0789343).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290228, 75751, 79711
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.