Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Importin 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111437 Supplier Invitrogen™ Supplier No. PA5111437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NRNPVWYQAL THGLNEEQRK QLQDIATLAD QRRAAHESKM IEKHGGYKFS APVVPSSFNF GGPAPGMN Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

The importin-alpha/beta complex and the GTPase Ran mediate nuclear import of proteins with a classical nuclear localization signal. The protein encoded by this gene is a member of a class of approximately 20 potential Ran targets that share a sequence motif related to the Ran-binding site of importin-beta. Similar to importin-beta, this protein prevents the activation of RaNULLs GTPase by RanGAP1 and inhibits nucleotide exchange on RanGTP, and also binds directly to nuclear pore complexes where it competes for binding sites with importin-beta and transportin. This protein has a Ran-dependent transport cycle and it can cross the nuclear envelope rapidly and in both directions. At least four importin beta-like transport receptors, namely importin beta itself, transportin, RanBP5 and RanBP7, directly bind and import ribosomal proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Importin 7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ipo7
Gene Accession No. O95373
Gene Alias A330055O14Rik; C330016G14; FLJ14581; Imp7; importin 7; importin-7; IPO7; MGC138673; RAN binding protein 7; RAN-binding protein 7; RanBP7
Gene Symbols Ipo7
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Importin 7. Recombinant protein control fragment (Product #RP-106714).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10527
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.