Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin 7 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501563318
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3318 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3318 Supplier Bethyl Laboratories, Inc Supplier No. A302726AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Importin-7 (IOP7) belongs to the Importin beta family and contains an importin N-terminal domain. It functions in nuclear protein import in association with importin-beta subunit KPNB1. It mediates autonomously the nuclear import of ribosomal proteins RPL23A, RPS7 and RPL5 and thenuclear import of H2A, H2B, H3 and H4 histones in vitro.Studies suggest itacts as a nuclear import receptor for the Glucocorticoid receptors. It also acts as a receptor for nuclear localization signals (NLS) and to promote translocation of import substrates through the nuclear pore complex (NPC) by an energy requiring, Ran-dependent mechanism.Reports indicatethat in HIV-1 infection,IOP7 binds and mediates the nuclear import of HIV-1 Rev.

Specifications

Antigen Importin 7
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene IPO7
Gene Accession No. O95373
Gene Alias FLJ14581, Imp7, MGC138673, RANBP7
Gene Symbols IPO7
Host Species Rabbit
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10527
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.