Learn More
Importin 7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302726A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Importin-7 (IOP7) belongs to the Importin beta family and contains an importin N-terminal domain. It functions in nuclear protein import in association with importin-beta subunit KPNB1. It mediates autonomously the nuclear import of ribosomal proteins RPL23A, RPS7 and RPL5 and thenuclear import of H2A, H2B, H3 and H4 histones in vitro.Studies suggest itacts as a nuclear import receptor for the Glucocorticoid receptors. It also acts as a receptor for nuclear localization signals (NLS) and to promote translocation of import substrates through the nuclear pore complex (NPC) by an energy requiring, Ran-dependent mechanism.Reports indicatethat in HIV-1 infection,IOP7 binds and mediates the nuclear import of HIV-1 Rev.Specifications
Importin 7 | |
Polyclonal | |
Unconjugated | |
IPO7 | |
FLJ14581, Imp7, MGC138673, RANBP7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10527 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95373 | |
IPO7 | |
Between 50 and 100 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.