Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Importin 8 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142011
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise isoform 1 (NP_006381.2) and isoform 2 (NP_001177924.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000. Preliminary analysis showed no signal in Western Blot.
Importin 8 is a 1037 amino acidnuclear protein with a RAN binding site. It belongs to RAN-binding protein super family and hence shares an N-terminal sequence motif with importin-beta that appears to account for RAN GTP binding. It binds to the nuclear pore complex (NPC) and is well known as a transport carrier that mediates nuclear import of ribosomal proteins with a classical nuclear localization signal. Importin 8 works by binding to the nuclear pore complex and, along with RANGTP and RANBP1, inhibits the GAP stimulation of the RAN GTPase. Along with transportin it also helps in the nuclear import of SRP19.
Specifications
Importin 8 | |
Polyclonal | |
Unconjugated | |
IPO8 | |
6230418K12Rik; Abcc10; ATP-binding cassette, sub-family C (CFTR/MRP), member 10; C130009K11Rik; FLJ26580; imp8; importin 8; importin-8; IPO8; MRP7; Om1; OM-1; oocyte maturation 1; RAN binding protein 8; ran-binding protein 8; RANBP8 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
10526 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O15397 | |
IPO8 | |
Peptide with sequence C-SAFNFGTVPSNN. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction