Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin-9 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen Importin-9
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056499T
SDP
View Documents
Novus Biologicals
NB10056499SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056499
SDP
View Documents
Novus Biologicals
NB10056499
0.1 mg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Importin-9 Polyclonal specifically detects Importin-9 in Human, Mouse, Rat, Canine, Primate, Rhesus Macaque samples. It is validated for Western Blot, Knockout Validated.
Specifications

Specifications

Importin-9
Western Blot 1-3 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Rat, Canine, Primate, Rhesus Macaque
Q96P70
55705
A portion of amino acids 850-950 of human Importin-9 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS containing 0.2% gelatin with 0.05% Sodium Azide
DKFZp761M1547, FLJ10402, Imp9IMP9, importin 9, importin-9, KIAA1192, Ran-binding protein 9, RANBP9
IPO9
IgG
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, _b, _c), mouse (isoforms 1 and 2), rat, dog and rhesus monkey, 88% homologous in chicken, 83% homologous in opossum, and 78% in bovine.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.