Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Importin-9 Antibody (SR1154), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322553100UL
Description
Importin-9 Monoclonal antibody specifically detects Importin-9 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Importin-9 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1154 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
DKFZp761M1547, FLJ10402, Imp9IMP9, importin 9, importin-9, KIAA1192, Ran-binding protein 9, RANBP9 | |
A synthesized peptide derived from human Importin-9 (Uniprot #: Q96P70) | |
100 μg | |
Core ESC Like Genes, Stem Cell Markers | |
55705 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction