Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Importin 9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675215
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-215 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-215 Supplier Invitrogen™ Supplier No. PA141598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: HeLa lysate. Chicken (82%), Rat, Bovine (82%).

Functions in nuclear protein import as nuclear transport receptor. Serves as receptor for nuclear localization signals (NLS) in cargo substrates. Is thought to mediate docking of the importin/substrate complex to the nuclear pore complex (NPC) through binding to nucleoporin and the complex is subsequently translocated through the pore by an energy requiring, Ran-dependent mechanism. At the nucleoplasmic side of the NPC, Ran binds to the importin, the importin/substrate complex dissociates and importin is re-exported from the nucleus to the cytoplasm where GTP hydrolysis releases Ran. The directionality of nuclear import is thought to be conferred by an asymmetric distribution of the GTP-and GDP-bound forms of Ran between the cytoplasm and nucleus. Mediates the nuclear import of H2B histone, RPS7 and RPL18A. Prevents the cytoplasmic aggregation of RPS7 and RPL18A by shielding exposed basic domains. May also import H2A, H3, H4 histones, RPL4 and RPL6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Importin 9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene IPO9
Gene Accession No. Q91YE6, Q96P70
Gene Alias 0710008K06Rik; AI845704; C78347; DKFZp761M1547; FLJ10402; HSPC273; Imp9; imp9a; imp9b; importin 9; importin-9; importin-9a; importin-9b; Ipo9; KIAA1192; mKIAA1192; Ran binding protein 9; ran-binding protein 9; RANBP9; RP11-90L20.4
Gene Symbols IPO9
Host Species Rabbit
Immunogen A portion of amino acids 950-1000 of human Importin-9.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226432, 55705, 707210
Target Species Human, Mouse, Monkey, Rhesus Macaque
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.