Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin beta/KPNB1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10094993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10094993 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10094993 Supplier Novus Biologicals Supplier No. NB10094993
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Importin beta/KPNB1 Polyclonal specifically detects Importin beta/KPNB1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Importin beta/KPNB1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q14974
Gene Alias IMB1, Impnb, importin 1, importin 90, importin beta-1 subunit, importin subunit beta-1, importin-90, IPO1, IPOB, karyopherin (importin) beta 1, Karyopherin subunit beta-1, MGC2155, MGC2156, MGC2157, NTF97Pore targeting complex 97 kDa subunit, Nuclear factor p97, PTAC97
Gene Symbols KPNB1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 826 and 876 of human karyopherin (importin) beta 1 using the numbering given in entry NP_002256.2 (GeneID 3837).
Molecular Weight of Antigen 97 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3837
Test Specificity Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rat, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Green anole, White-tufted-ear marmoset, and Naked mole rat.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.