Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IMUP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562962
Description
Immunogen sequence: SSSDSDTDVK SHAAGSKQHE SIPGKAKKPK VKKKEKGKK Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 62%.
IMUP is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosol; nucleus; plasma membrane.
Specifications
IMUP | |
Polyclonal | |
Unconjugated | |
C19orf33 | |
C19orf33; chromosome 19 open reading frame 33; H2RSP; HAI-2 related small protein; HAI-2-related small protein; hepatocyte growth factor activator inhibitor type 2-related small protein; Immortalization up-regulated protein; immortalization-upregulated protein; IMUP; IMUP-1; IMUP-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64073 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9GZP8 | |
C19orf33 | |
Recombinant protein corresponding to Human IMUP. Recombinant protein control fragment (Product #RP-100068). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction