Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INA Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501570408
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0408 Supplier Bethyl Laboratories, Inc Supplier No. A305431A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine

Alpha internexin is a Class IV intermediate filament originally discovered as it copurifies with other neurofilament subunits. On SDS-PAGE gels it runs with an apparent molecular weight of 64 to 66 kDa, with some species variability, although the real molecular weight is about 55kDa; as with the other neurofilament subunits the presence of highly negatively charged sequences results in reduction of SDS-PAGE mobility. a-internexin is related to but distinct from the better known neurofilament triplet proteins, NF-L, NF-M and NF-H, having similar protein sequence motifs and a similar intron organization. It is expressed only in neurons and in large amounts early in neuronal development, but is down-regulated in many neurons as development proceeds. Many classes of mature neurons contain a-internexin in addition to NF-L, NF-M and NF-H. In some mature neurons a-internexin is the only neurofilament subunit expressed. Antibodies to a-internexin are therefore unique probes to study and classify neuronal types and follow their processes in sections and in tissue culture. In addition, the very early developmental expression of a-internexin means its presence is an early and convenient diagnostic feature of neuronal progenitors cells and other cells committed to the neuronal lineage. In addition, recent studies show a marked up-regulation of a-internexin during neuronal regeneration.

Specifications

Antigen INA
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene INA
Gene Accession No. Q16352
Gene Alias 66 kDa neurofilament protein, Internexin neuronal intermediate filament protein alpha, MGC12702, NEF5, Neurofilament 5 (66kD), Neurofilament 66, Neuro
Gene Symbols INA
Host Species Rabbit
Immunogen Between 300 and 350
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9118
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.