Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INAVA Antibody (OTI6A6), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272021V
Description
INAVA Monoclonal specifically detects INAVA in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
INAVA | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C1ORF106 | |
Recombinant protein fragment corresponding to amino acids 1-270 of human C1orf106 (NP_060735) produced in E.coli. | |
0.1 mL | |
55765.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI6A6 | |
Western Blot, Immunohistochemistry | |
C1orf106, chromosome 1 open reading frame 106, hypothetical protein LOC55765 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction