Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INCA Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP146144
Description
INCA Polyclonal specifically detects INCA in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
INCA | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
caspase recruitment domain family, member 17, caspase recruitment domain-containing protein 17, Caspase-1 inhibitor INCA, INCAInhibitory CARD, inhibitory caspase recruitment domain (CARD) protein, Inhibitory caspase recruitment domain protein | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific to the C-Terminus. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:8000 | |
NP_001007233.1 | |
CARD17 | |
Peptide with sequence C-NHLTTQDSQIVLPS corresponding to C-Terminus according to NP_001007233.1. | |
0.1 mg | |
Apoptosis | |
440068 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction