Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INCA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560689
Description
Immunogen sequence: KKRRPCLEGM QQQGLGGVPA RVRAVTYHLE DLRRRQSIIN ELKKAQWGSS GAASEPVVLG EEGCGFPSTN EYPD Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 68%.
Binds to CDK2-bound cyclins and inhibits the kinase activity of CDK2; binding to cyclins is critical for its function as CDK inhibitor (PubMed:21540187). Inhibits cell growth and cell proliferation and may play a role in cell cycle control. Required for ING5-mediated regulation of S-phase progression, enhancement of Fas-induced apoptosis and inhibition of cell growth. [UniProt].
Specifications
INCA1 | |
Polyclonal | |
Unconjugated | |
INCA1 | |
AI842396; HSD45; INCA1; Inhibitor of CDK interacting with cyclin A1; inhibitor of CDK, cyclin A1 interacting protein 1; protein INCA1; RGD1306656 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0VD86 | |
INCA1 | |
Recombinant protein corresponding to Human INCA1. Recombinant protein control fragment (Product #RP-93541). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction