Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ INCB 024360-analog, >99%
GREENER_CHOICESDP

Catalog No. 600750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-075-0 50 mg
60-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-075-0 Supplier Tocris Bioscience™ Supplier No. 6007/50
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent indoleamine 2,3-dioxygenase (IDO) inhibitor

Potent indoleamine 2,3-dioxygenase (IDO) inhibitor (IC50 values are 19 and 67 nM in enzymatic activity and HeLa cell assays, respectively). Decreases kynurenine levels in plasma and reduces tumor growth in vivo. Cell membrane permeable.

Chemical Identifiers

CAS 914471-09-3
Molecular Formula C9H7ClFN5O2

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 271.64
Purity >99%
Molecular Weight (g/mol) 271.636
Chemical Name or Material 4-Amino-N'-(3-chloro-4-fluorophenyl)-N-hydroxy-1,2,5-oxadiazole-3-carboximidamide
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.