Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INCENP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517200 Supplier Invitrogen™ Supplier No. PA517200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Chromosomal passenger proteins are members of a group of proteins that move from centromeres to the spindle midzone during mitosis. INCENP (Inner Centromere Protein) is a chromosomal passenger protein with an essential role in mitosis and meiosis. It is a complex highly basic multidomain protein that is dynamically translocated from metaphase chromosomes to the spindle midzone during or just prior to anaphase. Some INCENP transfers to the equatorial cortex before formation of the cleavage furrow. Ultimately, it is discarded in the midbody at the completion of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INCENP
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 7.7 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Incenp
Gene Accession No. Q9NQS7
Gene Alias 2700067E22Rik; AU019509; binds and activates aurora-B and -C in vivo and in vitro; C130081E20; C77457; chromosomal passenger protein; Incenp; Inner centromere protein; inner centromere protein antigens 135/155kDa; inner centromere protein INCENP
Gene Symbols Incenp
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids surrounding Ala841 of human INCENP.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3619
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.