Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inconel∣r 600 gauze, 100 mesh woven from 0.102mm (0.004in) dia wire
Click to view available options
Quantity:
100 x 100 mm
150 x 150 mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Inconel™ 600 gauze |
| Name Note | 100 mesh woven from 0.102mm (0.004 in.) dia. wire |
| Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cr:Fe; 77:15.5:7.5 wt% |
| Quantity | 150 x 150 mm |
| Mesh Size | 100 Mesh |
| MDL Number | MFCD01741927 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction