Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heratherm Incubation Chamber

Catalog No. 01-672-442PM
Click to view available options
Door Style:
Glass
Solid Outer, Glass Inner

Catalog No. 01-672-442PM

Supplier: Thermo Scientific™ EIS750PM

Comprehensive data monitoring and temperature control combined with a flexible Heratherm Incubation Chamber provide a dependable option for microbiological incubation or temperature-dependent storage and testing.

  • Uniform Temperature throughout the chamber.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 26.5 cu.ft.
Capacity (English) Shelf 66.1 lb.
Capacity (Metric) Shelf 30 kg
Certifications/Compliance UL, CE
Data Outputs USB, Ethernet
Capacity (Metric) 750 L
Chamber Material Powder-coated Cold Rolled Steel
Lighting No
Material Powder-coated Cold Rolled Steel
No. of Shelves 3
Plug Type Nema 5-20P
Relative Humidity 10 to 90%
Temperature Range (Metric) 0 to 70°C
Type Environmental Chamber
Display Touchscreen
Door Style Solid Outer, Glass Inner
No. of Casters 4
Ports 2 Access Ports
Programmability Temperature Ramping
Resolution 0.1°C
Shelf Area (English) 4.9 ft.2
Shelf Area (Metric) 0.46 m2
Shelf Material Stainless Steel (304/1.4301)
Temperature Control Microprocessor
Temperature Stability ≤ ±0.1°C at 20°C to 37°C
Temperature Uniformity ≤ ±0.5°C at20°C to 37°C
Warranty 24 Months
Width (English) 42.52 in.
Width (English) Interior 33.07 in.
Width (English) Shelf 22.6 in.
Width (Metric) 108 cm
Width (Metric) Interior 84 cm
Width (Metric) Shelf 79.5 cm
Height (English) 72.24 in.
Height (English) Interior 57.09 in
Height (Metric) 183.5 cm
Height (Metric) Interior 145 cm
Depth (English) 34.84 in.
Depth (English) Interior 24.8 in.
Depth (Metric) 88.5 cm
Depth (Metric) Interior 63 cm
Shipping Weight (English) 840 lb.
Shipping Weight (Metric) 381 kg
Electrical Requirements 100/240 V, 50/60 Hz, 1400 W
Voltage 100/115/220/230 V
Wattage 900 W
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.