Learn More
OHAUS Incubator-Genie
Catalog No. 01362553 Shop All MilliporeSigma Supelco Products
Catalog No. 01-362-553 Supplier OHAUS Supplier No. CBL85
Only null left
Add to Cart
Add to Cart
Multi-functional shaking incubator that is also a hybridization oven.
  • Incorporates a magnetic platform and accessories.
  • Platform rocks and rotates.
  • Features a temperature controlled chamber ranging from 28 to 75°C.