Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ IND 1316
GREENER_CHOICESDP

Catalog No. 768550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-855-0 50 mg
76-851-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 76-855-0 Supplier Tocris Bioscience™ Supplier No. 7685/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AMPK activator

IND 1316 is an activator of AMP-activated protein kinase (AMPK). In vitro, it activates AMPK in a dose-dependent manner and induces different MAPK pathways (p38 and JNK). In vivo, it reduces the neuropathological symptoms in two animal models of Huntington's Disease (nematodes and mice). It shows very low toxicity even at high concentrations (>200 μM). This compound is orally bioavailable.

Specifications

Chemical Name or Material 3-[3-(Phenylmethoxy)phenyl]-1 H-indole-2-carboxylic acid
CAS 1888473-12-8
Quantity 50 mg
Target AMPK Activators
Molecular Formula C22H17NO3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.