Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indene, 90%, technical, stabilized

Catalog No. AC122140010
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle

CAS: 95-13-6 | C9H8 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-13-6, 98-29-3
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD00003777
InChI Key YBYIRNPNPLQARY-UHFFFAOYSA-N
Synonym indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy
PubChem CID 7219
ChEBI CHEBI:41921
IUPAC Name 1H-indene
SMILES C1C=CC2=CC=CC=C21

Specifications

Melting Point -5°C to 3°C
Color White to Yellow
Density 0.9900g/mL
Boiling Point 181.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5670 to 1.5720
Quantity 1 kg
Beilstein 05, 515
Specific Gravity 0.99
Merck Index 15, 4978
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents,soluble in acetone,miscible with ethanol and ethyl ether
Stabilizer 5 to 10ppm tert-butylcatechol
Formula Weight 116.16
Percent Purity 90%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Indene, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 202-393-6
RTECSNumber NK8225000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.