Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indene (Tech.), 90%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
Packaging:
Glass bottle
95-13-6,98-29-3
C9H8
116.16
MFCD00003777
YBYIRNPNPLQARY-UHFFFAOYSA-N
indene,indonaphthene,inden,unii-67h8y6lb8a,indenyl radical,1h-inden,pubchem20728,indene, reag,acmc-209ruy
7219
CHEBI:41921
1H-indene
C1C=CC2=CC=CC=C21
95-13-6,98-29-3
C9H8
116.16
MFCD00003777
YBYIRNPNPLQARY-UHFFFAOYSA-N
indene,indonaphthene,inden,unii-67h8y6lb8a,indenyl radical,1h-inden,pubchem20728,indene, reag,acmc-209ruy
7219
CHEBI:41921
1H-indene
C1C=CC2=CC=CC=C21
Melting Point -5°C to 3°C
Specific Gravity 0.99
Flash Point 56°C
Refractive Index 1.5670 to 1.5720
Stabilizer 5 to 10ppm tert-butylcatechol
Infrared Spectrum Authentic
Packaging Glass bottle
Color White to Yellow
Quantity 250g
Density 0.9900g/mL
Boiling Point 181.0°C
Assay Percent Range 90%
Beilstein 05, 515
Merck Index 15, 4978
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents,soluble in acetone,miscible with ethanol and ethyl ether
Formula Weight 116.16
Physical Form Crystalline Powder
Percent Purity 90%
Grade Technical
Chemical Name or Material Indene, tech., 90%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-393-6

RTECSNumber : NK8225000

TSCA : TSCA