Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indian Hedgehog/Ihh Antibody (9L16), Alexa Fluor™ 750, Novus Biologicals™
SDP

Rat Monoclonal Antibody

Supplier:  Novus Biologicals NBP222020AF750

Catalog No. NP222020A70


Only null left
Add to Cart

Description

Description

Indian Hedgehog/Ihh Monoclonal specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Indian Hedgehog/Ihh
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
IHH
Human recombinant Indian Hedgehog/Ihh (N-terminus)
Primary
Human
IgG2
Western Blot, ELISA
9L16
Western Blot, ELISA
BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein
Rat
0.1 mL
3549
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.