Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indian Hedgehog/Ihh Antibody (9L16) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019288 0.025 mg
NBP222020 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019288 Supplier Novus Biologicals Supplier No. NBP2220200.025MG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Indian Hedgehog/Ihh Monoclonal specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Indian Hedgehog/Ihh
Applications Western Blot, ELISA
Classification Monoclonal
Clone 9L16
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:2000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. NP_002172
Gene Alias BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein
Gene Symbols IHH
Host Species Rat
Immunogen This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human Indian Hedgehog Nterminus. Rat anti human Indian Hedgehog N-Term NP_002172.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3549
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.