Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indian Hedgehog/Ihh Antibody (9L16), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP222020G
Description
Indian Hedgehog/Ihh Monoclonal specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot, ELISA.Specifications
Indian Hedgehog/Ihh | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IHH | |
Human recombinant Indian Hedgehog/Ihh (N-terminus) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
9L16 | |
Western Blot, ELISA | |
BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein | |
Rat | |
Protein G purified | |
Stem Cell Signaling Pathway | |
3549 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction