Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indicator 1 (Bromophenol Blue), Reagents
Click to view available options
Quantity:
120 mL
1 L
Specifications
Specifications
| Chemical Name or Material | Indicator 1 (Bromophenol Blue) |
| CAS | 7732-18-5, 115-39-9 |
| Boiling Point | 100°C |
| Synonym | 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0004 |
| Quantity | 120 mL |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction