Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indicator 4 (Ferroin), Reagents
Click to view available options
Quantity:
100 mL
Specifications
Specifications
Chemical Name or Material | Indicator 4 (Ferroin) |
CAS | 7732-18-5, 5144-89-8 |
Boiling Point | 100°C |
Synonym | o-Phenanthroline, 1,10-Phenanthroline |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.01 |
Color | Red |
Quantity | 100 mL |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction