Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indigo Carmine (Acid Blue 74), Reagents

Catalog No. p-200022027
Click to view available options
Quantity:
10 g
25 g

Catalog No. 01-336-752

Supplier: Reagents Holdings Llc C611400010C1

Only null left
Add to Cart
Add to Cart

Indigo Carmine (Acid Blue 74), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Indigo Carmine (Acid Blue 74)
Boiling Point 300°C
CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Synonym FD and C Blue 2
Molecular Weight (g/mol) 466.3529
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.01
Color Blue
Quantity 10 g
Physical Form Solid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.