Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Indiko™ Plus Clinical Chemistry Analyzer

Catalog No. 23-278-456
Click to view available options
Description:
Indiko Plus without ISE Unit

Experience an uncompromised level of performance and ease of use with the Thermo Scientific™ Indiko™ Plus Clinical Chemistry Analyzer.

Experience an uncompromised level of performance and ease of use with the Thermo Scientific™ Indiko™ Plus Clinical Chemistry Analyzer. This fully-automated random access benchtop system for clinical and specialty chemistry testing, combined with system reagents and consumables form a fully-supported, complete system solution offering confidence to daily work.

  • IVDR-compliant, cost-effective solution for small and medium laboratory settings
  • Methods used in Indiko Plus are traceable to international reference methods and materials
  • Sample oriented testing generates results quickly, enhancing the quality of patient care
  • Application protocols include routine clinical chemistry tests as well as special tests, such as specific proteins, drug of abuse testing, therapeutic drug monitoring and immunosuppressant drug testing
  • Simple and quick daily start-up and maintenance procedures
  • Continuous access to samples, cuvettes and reagents without interrupting the testing process
  • Disposable cuvettes allow user-safe and contamination free analysis
  • Versatile, automated dilution features make sample processing easier
  • Reflex testing capability offers savings in operator time, triggering automatically the specific test to be run
  • Measurement reliability can be monitored with a real-time QC program
  • Automatic upload of calibrator and control values save time and reduce manual errors
  • Results archiving associated with calibration and reagent lot data secure full traceability
TRUSTED_SUSTAINABILITY

Specifications

Type Clinical Chemistry Analyzer
Description Indiko Plus without ISE Unit
Analytical Method Clinical and Specialty Chemistry Testing with Random Access Analysis
Capacity Capacity: photometric tests/hr: up to 350; On-board capacity: up to 108 sample positions and 42 reagent positions
Cuvettes Disposable Cuvette
Distilled Water Consumption 2.5 L/hr.
Reagents Reagent volume: 2 to 240µL (1 to 4 reagent additions/test)
Quantity Each
Height (English) 24.4 in.
Height (Metric) 62 cm
Width (English) 37 in.
Width (Metric) 94 cm
Weight (English) 242 lb.
Weight (Metric) 110 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.