Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Indinavir sulfate
GREENER_CHOICESDP

Catalog No. 719650 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-965-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 71-965-0 Supplier R&D Systems™ Supplier No. 7196/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective HIV-1 and HIV-2 protease inhibitor

Indinavir sulfate is a potent and selective HIV protease inhibitor (Ki values are 0.52 and 3.3 nM for HIV-1 and HIV-2, respectively). Indinavir stops the spread of the HIV-infected MT4 lymphoid cells at concentrations of 25-50 nM. Indinavir disrupts the ERK1/2 and p38 MAPK pathways and reduces both the level of functional eIF4F complex and rpS6 phosphorylation, which leads to a decrease in protein synthesis in mouse myocytes. In silico docking models predict potential as an inhibitor of SARS-CoV-2 3CLpro.

Specifications

Chemical Name or Material 2,3,5-Trideoxy-N-[(1 S,2 R)-2,3-dihydro-2-hydroxy-1 H-inden-1-yl]-5-[(2 S)-2-[[(1,1-dimethylethyl)amino]carbonyl]-4-(3-pyridinylmethyl)-1-piperazinyl]-2-(phenylmethyl)-D-erythro-pentonamide sulfate
CAS 157810-81-6
Quantity 50 mg
Product Line Tocris
Target HIV Protease Inhibitors
Molecular Formula C36H47N5O4.H2SO4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.