Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indinavir sulfate hydrate, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Color | White to Light Brown |
Description | Indinavir sulfate hydrate |
Molecular Formula | C36 H47 N5 O4 . H2 S O4 . x H2 O |
Shipping Condition | No special requirements |
Product Line | Thermo Scientific™ |
Quantity | 5 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction