Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Indium
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Description
Indium
Specifications
Specifications
| CAS | 7440-74-6 |
| Melting Point | 156.6°C |
| Density | 7.3 g/mL (at 25°C) |
| Molecular Formula | In |
| Linear Formula | In |
| MDL Number | MFCD00134048 |
| Quantity | 1 Ea. |
| Synonym | Indium; IN000266 |
| Molecular Weight (g/mol) | 114.82 |
| Percent Purity | 100.0% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction