Alfa Aesar™ Indium antimonide, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1462604

Quantity 2g

 View more versions of this product

Catalog No. AA1462604

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1312-41-0
    Formula Weight 236.57
    MDL Number MFCD00016146
    UN Number UN1549
    Merck Index 14,4948
    Chemical Name or Material Indium antimonide

    Specifications

    Formula Weight 236.57
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Physical Form Pieces
    MDL Number MFCD00016146
    CAS 1312-41-0
    Melting Point 535°C
    Health Hazard 1 Warning
    TSCA TSCA
    Particle Size 6mm & down
    UN Number UN1549
    Merck Index 14,4948
    Quantity 2g
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Density 5.75g/mL at m.p.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
    Chemical Name or Material Indium antimonide
    Assay 99.999%, (metals basis)

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit