Learn More
Indium antimonide, Electronic Grade, 99.99% (metals basis)
Description
Indium antimonide is used in semiconductor electronics, as an infrared detector and filter. Used in thermal imaging cameras, FLIR systems, infrared homing missile guidance systems, photo-Dember emitter and in infrared astronomy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 535°C |
| Physical Form | Pieces |
| Quantity | 50 g |
| UN Number | UN1549 |
| Merck Index | 14,4948 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 236.57 |
| Percent Purity | 99.99% |
| Grade | Electronic |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H302+H332 |
| Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S. |
| EINECSNumber | 215-192-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.