Learn More
Indium arsenide, 99.9999% (metals basis)
Description
Indium arsenide is used for construction of infrared detectors, for the wavelength range of 1-3μm. It is widely used as terahertz radiation source as it is a strong photo-Dember emitter. Indium arsenide is also used for making of diode laser.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 936°C |
Color | Gray |
Physical Form | Pieces |
Quantity | 50 g |
Assay Percent Range | 99.9999% (Metal basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 189.74 |
Odor | Odorless |
Appearance | Gray polycrystalline pieces |
Chemical Name or Material | Indium arsenide, Hardly attacked by mineral acids |
Safety and Handling
Hazard Category | H301+H331-H350 |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 215-115-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.