Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium arsenide, 99% (metals basis)

Catalog No. p-7049779
Click to view available options
Quantity:
2 g
10 g

In semiconductor electronics | CAS: 1303-11-3 | AsIn | 189.74 g/mol

Indium arsenide is used in semiconductor electronics, construction of infrared detectors, terahertz radiation source, as it is a strong Photo-dember emitter, superior electron mobility and velocity, in high-power applications detector, diode lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-11-3
Molecular Formula AsIn
Molecular Weight (g/mol) 189.74
MDL Number MFCD00016144
InChI Key RPQDHPTXJYYUPQ-UHFFFAOYSA-N
Synonym indium arsenide, indiam arsenide, indium monoarsenide, indium arsenide inas, arsanylidyneindium, dsstox_cid_3825, dsstox_rid_77199, dsstox_gsid_23825, indium arsenide 1g
PubChem CID 91500
IUPAC Name indiganylidynearsane
SMILES [As]#[In]

Specifications

Melting Point 936°C
Physical Form Pieces
Quantity 2 g
Linear Formula InAs
UN Number UN1557
Merck Index 14,4949
Solubility Information Insoluble in water.
Formula Weight 189.74
Percent Purity 99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Indium arsenide
Show More Show Less
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 215-115-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.