Learn More
Indium arsenide, 99% (metals basis), Thermo Scientific Chemicals
Description
Indium arsenide is used in semiconductor electronics, construction of infrared detectors, terahertz radiation source, as it is a strong Photo-dember emitter, superior electron mobility and velocity, in high-power applications detector, diode lasers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 936°C |
Physical Form | Pieces |
Quantity | 2 g |
Linear Formula | InAs |
UN Number | UN1557 |
Merck Index | 14,4949 |
Solubility Information | Insoluble in water. |
Formula Weight | 189.74 |
Percent Purity | 99% |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H301+H331-H350 |
Hazard Statement | GHS H Statement H301-H331 Toxic if swallowed. Toxic if inhaled. |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S. |
EINECSNumber | 215-115-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.